Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655053.1 | XP_007653243.1 | 20 - 21 | 4086 | 99 | |
GenBank mRNA | JI891679.1 | - | 5 - 6 | 1123 | 99 | |
JP037827.1 | - | 5 - 6 | 1122 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |