Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655053.1 | XP_007653243.1 | 23 - 24 | 4338 | 84 | |
GenBank mRNA | JI891679.1 | - | 8 - 9 | 1375 | 84 | |
JP037827.1 | - | 8 - 9 | 1374 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |