Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500660.2 | XP_003500708.1 | 2 - 3 | 913 | 96 | |
XM_007641413.1 | XP_007639603.1 | 3 - 4 | 669 | 96 | ||
XM_007641414.1 | XP_007639604.1 | 3 - 4 | 697 | 96 |