Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511521.2 | XP_003511569.2 | 1 - 2 | 67 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |