Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510698.2 | XP_003510746.1 | 4 - 5 | 481 | 89 | |
GenBank mRNA | JP042038.1 | - | 4 - 5 | 469 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |