Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653057.1 | XP_007651247.1 | 2 - 3 | 175 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |