Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653057.1 | XP_007651247.1 | 3 - 4 | 298 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |