Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513019.2 | XP_003513067.1 | 4 - 5 | 1826 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Probe spans exons |