Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648251.1 | XP_007646441.1 | 3 - 4 | 328 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |