Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506417.1 | XP_003506465.1 | 1 - 2 | 211 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Probe spans exons |