Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502048.2 | XP_003502096.2 | 2 - 3 | 396 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |