Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652380.1 | XP_007650570.1 | 2 - 3 | 505 | 103 | |
XM_007652381.1 | XP_007650571.1 | 2 - 3 | 505 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |