Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam72a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501214.2 | XP_003501262.1 | 1 - 2 | 991 | 103 | |
GenBank mRNA | JI871624.1 | - | 1 - 2 | 330 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |