Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502191.2 | XP_003502239.1 | 1 - 2 | 241 | 95 | |
GenBank mRNA | JI870939.1 | - | 1 - 2 | 226 | 95 | |
JP047507.1 | - | 1 - 2 | 227 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |