Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509378.1 | XP_003509426.1 | 5 - 6 | 1177 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |