Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642885.1 | XP_007641075.1 | 1 - 2 | 110 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |