Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500947.2 | XP_003500995.1 | 2 - 3 | 574 | 104 | |
GenBank mRNA | JP048356.1 | - | 2 - 3 | 561 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |