Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502727.2 | XP_003502775.1 | 2 - 3 | 306 | 72 | |
GenBank mRNA | JI889944.1 | - | 1 - 2 | 38 | 72 | |
JP050944.1 | - | 2 - 3 | 212 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Probe spans exons |