Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510485.2 | XP_003510533.1 | 2 - 3 | 745 | 84 | |
GenBank mRNA | JI890520.1 | - | 2 - 3 | 288 | 84 | |
JP046550.1 | - | 2 - 3 | 288 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |