Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sohlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653206.1 | XP_007651396.1 | 4 - 5 | 440 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |