Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644384.1 | XP_007642574.1 | 3 - 4 | 514 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Probe spans exons |