Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498994.2 | XP_003499042.1 | 1 | 406 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Both primers and probe map within a single exon |