Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503735.1 | XP_003503783.1 | 1 - 2 | 363 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |