Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgfrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641376.1 | XP_007639566.1 | 3 - 4 | 854 | 51 | |
GenBank mRNA | JI878885.1 | - | 1 - 2 | 354 | 51 | |
JP038778.1 | - | 1 - 2 | 354 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |