Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504653.2 | XP_003504701.1 | 2 - 3 | 565 | 68 | |
GenBank mRNA | JP046225.1 | - | 1 - 2 | 72 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |