Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502819.2 | XP_003502867.1 | 2 - 3 | 414 | 69 | |
GenBank mRNA | JI867497.1 | - | 2 - 3 | 237 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |