Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tppp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513949.1 | XP_003513997.1 | 1 - 2 | 315 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |