Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643088.1 | XP_007641278.1 | 2 - 3 | 527 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |