Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497198.2 | XP_003497246.1 | 1 - 2 | 167 | 91 | |
GenBank mRNA | JI864718.1 | - | 1 - 2 | 161 | 91 | |
JP045380.1 | - | 1 - 2 | 161 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |