Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502462.2 | XP_003502510.1 | 4 - 5 | 1209 | 66 | |
XM_007643077.1 | XP_007641267.1 | 4 - 5 | 1306 | 66 | ||
GenBank mRNA | JI864234.1 | - | 4 - 5 | 1192 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |