Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007626360.1 | XP_007624550.1 | 1 - 2 | 73 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |