Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497679.2 | XP_003497727.2 | 3 - 4 | 489 | 67 | |
GenBank mRNA | JI875976.1 | - | 3 - 4 | 489 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |