Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007608718.1 | XP_007606908.1 | 3 - 4 | 396 | 91 | |
GenBank mRNA | JI888463.1 | - | 3 - 4 | 396 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |