Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650545.1 | XP_007648735.1 | 1 - 2 | 253 | 65 | |
XM_007650546.1 | XP_007648736.1 | 1 - 2 | 253 | 65 | ||
XM_007650547.1 | XP_007648737.1 | 1 - 2 | 253 | 65 | ||
XM_007650548.1 | XP_007648738.1 | 1 - 2 | 253 | 65 | ||
GenBank mRNA | JI880802.1 | - | 1 - 2 | 238 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |