Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnmtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506719.2 | XP_003506767.1 | 1 - 2 | 360 | 78 | |
GenBank mRNA | JI883449.1 | - | 1 - 2 | 327 | 78 | |
JP062970.1 | - | 1 - 2 | 327 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |