Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501210.2 | XP_003501258.1 | 4 - 5 | 552 | 82 | |
XM_007641901.1 | XP_007640091.1 | 5 - 6 | 643 | 82 | ||
GenBank mRNA | JI866677.1 | - | 2 - 3 | 307 | 82 | |
JP063185.1 | - | 2 - 3 | 307 | 82 |