Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652289.1 | XP_007650479.1 | 5 - 6 | 656 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |