Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499913.2 | XP_003499961.1 | 1 - 2 | 346 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |