Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503135.2 | XP_003503183.1 | 4 - 5 | 484 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |