Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503568.1 | XP_003503616.1 | 1 - 2 | 331 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |