Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513430.2 | XP_003513478.1 | 5 - 6 | 840 | 67 | |
GenBank mRNA | JI896345.1 | - | 2 - 3 | 369 | 67 | |
JP054966.1 | - | 2 - 3 | 369 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |