Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512368.2 | XP_003512416.1 | 3 - 4 | 430 | 75 | |
XM_007652860.1 | XP_007651050.1 | 3 - 4 | 431 | 75 | ||
GenBank mRNA | JI875984.1 | - | 3 - 4 | 365 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |