Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651221.1 | XP_007649411.1 | 1 - 2 | 602 | 58 | |
GenBank mRNA | JI893212.1 | - | 1 - 2 | 603 | 58 | |
JP060173.1 | - | 1 - 2 | 602 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |