Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007629274.1 | XP_007627464.1 | 4 - 5 | 1395 | 58 | |
XM_007629280.1 | XP_007627470.1 | 3 - 4 | 1242 | 58 | ||
XM_007629284.1 | XP_007627474.1 | 4 - 5 | 1334 | 58 | ||
XM_007629291.1 | XP_007627481.1 | 4 - 5 | 916 | 58 | ||
XM_007629298.1 | XP_007627488.1 | 2 - 3 | 536 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |