Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499194.2 | XP_003499242.1 | 5 - 6 | 460 | 81 | |
GenBank mRNA | JI896416.1 | - | 2 - 3 | 264 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |