Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100768630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504396.2 | XP_003504444.1 | 15 - 16 | 1942 | 136 | |
XM_007644919.1 | XP_007643109.1 | 15 - 16 | 1942 | 136 | ||
XM_007644920.1 | XP_007643110.1 | 15 - 16 | 1942 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |