Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100768630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504396.2 | XP_003504444.1 | 22 - 23 | 2820 | 105 | |
XM_007644919.1 | XP_007643109.1 | 22 - 23 | 2820 | 105 | ||
GenBank mRNA | JP042185.1 | - | 2 - 3 | 206 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |