Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505922.2 | XP_003505970.1 | 2 - 3 | 317 | 76 | |
GenBank mRNA | JI896513.1 | - | 1 - 2 | 154 | 76 | |
JP037634.1 | - | 1 - 2 | 154 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |