Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497497.2 | XP_003497545.1 | 3 - 4 | 699 | 86 | |
XM_007613105.1 | XP_007611295.1 | 3 - 4 | 852 | 86 | ||
XM_007613121.1 | XP_007611311.1 | 3 - 4 | 854 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |