Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rergl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507097.2 | XP_003507145.1 | 2 - 3 | 101 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |